Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021391.3 | NP_067366.1 | 4 | 349 | ||
GenBank mRNA | AF281676.1 | - | 4 | 239 | ||
BC025123.1 | - | 4 | 318 | 4019442 | ||
CA321941.1 | - | 4 | 349 |