Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190984.1 | NP_001177913.1 | 10 | 1500 | ||
NM_001190985.1 | NP_001177914.1 | 9 | 1718 | |||
NM_021295.3 | NP_067270.1 | 10 | 1534 | |||
GenBank mRNA | AJ294535.1 | - | 10 | 1527 |