Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021303.2 | NP_067278.2 | 5 | 561 | ||
GenBank mRNA | AK076287.1 | - | 4 | 603 | ||
AK078560.1 | - | 5 | 537 | |||
AK133834.1 | - | 5 | 561 | |||
AK157765.1 | - | 5 | 553 | |||
AK164071.1 | - | 1 | 1478 | |||
BC020013.1 | - | 5 | 529 | 3663482 |