Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021430.2 | NP_067405.1 | 1 | 688 | ||
GenBank mRNA | AB041584.1 | - | 1 | 318 | ||
BC029965.1 | - | 1 | 366 | |||
BC051945.1 | - | 1 | 688 | 5716540 | ||
BC051946.1 | - | 1 | 618 | 6403995 | ||
BC058384.1 | - | 1 | 581 | 5718399 |