Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021509.5 | NP_067484.2 | 1 | 198 | ||
GenBank mRNA | AB041606.1 | - | 1 | 133 | ||
AK014340.1 | - | 1 | 199 | |||
AK030274.1 | - | 1 | 139 | |||
AK081586.1 | - | 1 | 164 | |||
AK162030.1 | - | 1 | 140 | |||
BC037684.1 | - | 1 | 136 | |||
BC057652.1 | - | 1 | 112 | 4017199 |