Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021448.2 | NP_067423.2 | 3, 4 | 330 | ||
GenBank mRNA | AF291064.1 | - | 3, 4 | 296 | ||
AK033246.1 | - | 3, 4 | 331 | |||
AK034457.1 | - | 3, 4 | 329 | |||
AK044699.1 | - | 3, 4 | 307 | |||
AK050960.1 | - | 3, 4 | 331 | |||
AK147826.1 | - | 331 | ||||
AK167725.1 | - | 3, 4 | 329 | |||
AK167853.1 | - | 3, 4 | 329 | |||
AK168370.1 | - | 3, 4 | 331 | |||
AK177305.1 | - | 1, 2 | 95 | |||
BC015284.1 | - | 3, 4 | 322 | 4239056 | ||
BC051943.1 | - | 3, 4 | 322 | 5686798 |