Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 2, 3 | 265 | ||
GenBank mRNA | AF263247.1 | - | 2, 3 | 227 | ||
AK007307.1 | - | 2, 3 | 236 | |||
AK034859.1 | - | 2, 3 | 264 | |||
AK049424.1 | - | 2, 3 | 285 | |||
AK075637.1 | - | 1, 2 | 105 | |||
AK088854.1 | - | 2, 3 | 273 | |||
AK145800.1 | - | 2, 3 | 328 | |||
AK152523.1 | - | 2, 3 | 273 | |||
AK168788.1 | - | 2, 3 | 291 | |||
AK169149.1 | - | 2, 3 | 273 | |||
BC039054.1 | - | 2, 3 | 251 | 2648667 | ||
BC058687.1 | - | 2, 3 | 263 | 6411707 | ||
CX212584.1 | - | 2, 3 | 265 |