Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021502.2 | NP_067477.1 | 2, 3 | 247 | ||
GenBank mRNA | AB030200.1 | - | 2, 3 | 240 | ||
AK007526.1 | - | 2, 3 | 217 | |||
BC028502.1 | - | 2, 3 | 227 | 1366538 | ||
BC114968.1 | - | 215 | ||||
BY091059.1 | - | 2, 3 | 248 |