Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021434.5 | NP_067409.2 | 9 | 1609 | ||
GenBank mRNA | AB041545.1 | - | 9 | 1601 | ||
AK040165.1 | - | 3 | 505 | |||
AK053422.1 | - | 9 | 1579 | |||
AK169908.1 | - | 9 | 1608 | |||
AK170316.1 | - | 9 | 1574 | |||
BC055726.1 | - | 9 | 1584 | |||
BC069981.1 | - | 9 | 1565 | 6336270 |