Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021491.3 | NP_067466.1 | 8, 9 | 2346 | ||
GenBank mRNA | AJ250461.1 | - | 6, 7 | 1877 | ||
AK019476.2 | - | 9, 10 | 2398 | |||
AK076416.1 | - | 8, 9 | 2323 | |||
AK139226.1 | - | 8, 9 | 2322 | |||
AK140788.1 | - | 4, 5 | 348 | |||
BC043077.1 | - | 8, 9 | 2346 | 5721118 | ||
BC046980.1 | - | 7, 8 | 2166 | 6399438 |