Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146001.1 | NP_001139473.1 | 2 | 140 | ||
NM_021517.2 | NP_067492.2 | 2 | 186 | |||
GenBank mRNA | AF220100.1 | - | 2 | 125 | ||
AF474247.1 | - | 1 | 124 | |||
AK006269.1 | - | 2 | 660 | |||
AK012070.1 | - | 2 | 176 | |||
AK014879.1 | - | 2 | 443 | |||
AK029752.1 | - | 2 | 644 | |||
AK029764.1 | - | 2 | 654 | |||
AK165415.1 | - | 2 | 121 | |||
AW259368.1 | - | 2 | 186 | |||
BC013512.1 | - | 1 | 109 | 4234949 |