Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042726.3 | NP_001036191.1 | 1 | 571 | ||
NM_021543.4 | NP_067518.2 | 1 | 571 | |||
GenBank mRNA | AF231125.1 | - | 1 | 492 | ||
AK048638.1 | - | 1 | 572 | |||
AK049982.1 | - | 1 | 574 | |||
AK140407.1 | - | 1 | 456 | |||
BC052388.1 | - | 1 | 552 | 5718846 | ||
BC053008.1 | - | 1 | 559 | 5698009 |