Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021552.5 | NP_067527.3 | 4 | 531 | ||
GenBank mRNA | AF238866.1 | - | 4 | 457 | ||
AK002474.1 | - | 3 | 382 | |||
AK012832.1 | - | 4 | 472 | |||
AK014275.1 | - | 4 | 508 | |||
AK017605.1 | - | 4 | 472 | |||
AK077696.1 | - | 473 | ||||
AK078457.1 | - | 4 | 484 | |||
BC086776.1 | - | 3 | 327 | |||
BC108365.1 | - | 4 | 457 | 4219428 | ||
BC116384.1 | - | 4 | 472 | |||
BC126976.1 | - | 4 | 471 | 40101726 | ||
BC172160.1 | - | 4 | 445 |