Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021885.4 | NP_068685.1 | 4 | 604 | ||
GenBank mRNA | AK147700.1 | - | 4 | 659 | ||
AK165720.1 | - | 4 | 605 | |||
BC096554.1 | - | 4 | 585 | 30616697 | ||
U52433.1 | - | 4 | 592 | |||
U52824.1 | - | 5 | 737 | |||
U54643.1 | - | 4 | 522 |