Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021607.3 | NP_067620.3 | 4 | 408 | ||
GenBank mRNA | AF240469.1 | - | 4 | 358 | ||
AK033314.1 | - | 4 | 350 | |||
AK129102.1 | - | 4 | 408 | |||
AK170173.1 | - | 4 | 354 | |||
AK170244.1 | - | 4 | 373 | |||
AK170369.1 | - | 4 | 397 | |||
AK172579.1 | - | 4 | 353 | |||
BC019998.1 | - | 4 | 353 | 4458780 | ||
CD549803.1 | - | 4 | 408 |