Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021885.4 | NP_068685.1 | 4, 5 | 613 | ||
GenBank mRNA | AK147700.1 | - | 4, 5 | 668 | ||
AK165720.1 | - | 4, 5 | 614 | |||
BC096554.1 | - | 4, 5 | 594 | 30616697 | ||
U52433.1 | - | 4, 5 | 601 | |||
U52824.1 | - | 5, 6 | 746 | |||
U54643.1 | - | 4, 5 | 531 |