Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163354.1 | NP_001156826.1 | 15 | 1448 | ||
NM_001163355.1 | NP_001156827.1 | 15 | 1448 | |||
NM_021536.7 | NP_067511.4 | 15 | 1448 | |||
GenBank mRNA | AK019059.1 | - | 15 | 1408 | ||
AK028597.1 | - | 15 | 1391 | |||
AK031139.1 | - | 13 | 1110 | |||
AK032096.1 | - | 15 | 1421 | |||
AK041119.1 | - | 15 | 1384 | |||
AK043323.1 | - | 6 | 505 | |||
AK054472.1 | - | 15 | 1419 | |||
AK078955.1 | - | 15 | 1391 | |||
AK090071.1 | - | 8 | 774 | |||
AK138830.1 | - | 15 | 1389 | |||
AK141621.1 | - | 15 | 1275 | |||
AK147749.1 | - | 15 | 1427 | |||
AK147801.1 | - | 15 | 1453 | |||
AK167502.1 | - | 15 | 1448 | |||
AK169811.1 | - | 14 | 2105 | |||
BC006918.1 | - | 6 | 539 | |||
BC025021.1 | - | 1 | 561 | |||
BC044301.1 | - | 4 | 287 | |||
BC046785.1 | - | 15 | 1431 | 6416127 | ||
BC052882.1 | - | 5 | 392 | |||
BC058350.1 | - | 15 | 1363 | 6815832 |