Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021885.4 | NP_068685.1 | 8 | 1131 | ||
GenBank mRNA | AK147700.1 | - | 8 | 1186 | ||
BC096554.1 | - | 8 | 1112 | 30616697 | ||
U52433.1 | - | 8 | 1119 | |||
U52824.1 | - | 9 | 1264 | |||
U54643.1 | - | 8 | 1049 |