Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022023.2 | NP_071306.2 | 3, 4 | 297 | ||
GenBank mRNA | AB001732.1 | - | 3, 4 | 229 | ||
AB050013.1 | - | 3, 4 | 239 | |||
AF297220.1 | - | 3, 4 | 275 | |||
AK013947.1 | - | 3, 4 | 254 | |||
AK013970.1 | - | 3, 4 | 250 | |||
AK142014.1 | - | 3, 4 | 269 | |||
BC040233.1 | - | 3, 4 | 231 | 5364203 | ||
BC049134.1 | - | 3, 4 | 235 | 6831501 |