Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Popdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF204175.1 | - | 1 | 499 | ||
AK084713.1 | - | 1 | 531 | |||
BC064005.1 | - | 1 | 535 | 30287674 | ||
CB590067.1 | - | 1 | 534 |