Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258459.1 | NP_001245388.1 | 4 | 476 | ||
NM_001258460.1 | NP_001245389.1 | 4 | 449 | |||
NM_006213.4 | NP_006204.1 | 4 | 476 | |||
XM_017012324.1 | XP_016867813.1 | 4 | 476 | |||
GenBank mRNA | AK293180.1 | - | 4 | 431 | ||
AK300928.1 | - | 4 | 401 | |||
AK312851.1 | - | 4 | 476 | |||
BC051327.1 | - | 4 | 414 | 6277230 | ||
BC069655.1 | - | 4 | 392 | 7262499 | ||
BC069679.1 | - | 4 | 392 | 7262511 | ||
BC069738.1 | - | 4 | 393 | 7262523 | ||
BC069754.1 | - | 4 | 393 | 7262535 | ||
BC074753.2 | - | 3 | 320 | 30915212 | ||
DB171823.1 | - | 4 | 476 | |||
X80590.1 | - | 4 | 400 |