Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gmfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039192.1 | NP_001034281.1 | 7 | 576 | ||
NM_001290550.1 | NP_001277479.1 | 6 | 502 | |||
NM_022024.2 | NP_071307.1 | 7 | 599 | |||
GenBank mRNA | AB003588.1 | - | 7 | 533 | ||
AF297221.1 | - | 7 | 596 | |||
AK002834.1 | - | 7 | 600 | |||
AK009967.1 | - | 7 | 576 | |||
AK014214.1 | - | 7 | 600 | |||
AK152486.1 | - | 4 | 438 | |||
AK153228.1 | - | 4 | 438 | |||
BC011488.1 | - | 7 | 552 | 3581858 | ||
BE629139.1 | - | 6 | 462 |