Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022419.3 | NP_071864.2 | 4 | 1144 | ||
GenBank mRNA | AB030191.1 | - | 4 | 1134 | ||
AK003090.1 | - | 4 | 1132 | |||
AK080618.1 | - | 4 | 1143 | |||
AK155503.1 | - | 4 | 1144 | |||
AK181236.1 | - | 1 | 41 | |||
BC026540.1 | - | 4 | 1093 | 5050705 |