Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022654.1 | NP_073145.1 | 12 | 2207 | ||
GenBank mRNA | AF274973.1 | - | 12 | 2207 | ||
AK004702.1 | - | 12 | 2227 | |||
AK087880.1 | - | 2569 | ||||
AK156863.1 | - | 14 | 2195 | |||
BC032202.1 | - | 12 | 2084 | |||
BC056987.1 | - | 12 | 2558 | |||
BC089373.1 | - | 12 | 2179 | |||
BC099686.1 | - | 12 | 2545 | |||
BC145314.1 | - | 12 | 2211 | |||
BC145857.1 | - | 12 | 2172 | 40131160 |