Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023041.3 | NP_075528.3 | 2, 3 | 198 | ||
GenBank mRNA | AK012785.1 | - | 2, 3 | 181 | ||
AK029368.1 | - | 2, 3 | 199 | |||
AK075576.1 | - | 2, 3 | 185 | |||
AK170175.1 | - | 2, 3 | 187 | |||
BC012517.1 | - | 2, 3 | 133 | |||
BC019767.1 | - | 2, 3 | 181 | 4502819 | ||
CT010305.1 | - | 2, 3 | 174 | |||
Y09046.1 | - | 2, 3 | 179 |