Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023113.5 | NP_075602.2 | 1 | 262 | ||
GenBank mRNA | AF212998.1 | - | 1 | 112 | ||
AI527854.1 | - | 1 | 262 | |||
AK002859.1 | - | 1 | 260 | |||
AK008354.1 | - | 1 | 265 | |||
BC024934.1 | - | 1 | 226 | 4012726 |