Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023143.3 | NP_075632.3 | 2 | 391 | ||
GenBank mRNA | AF148216.1 | - | 2 | 379 | ||
AI119268.1 | - | 2 | 367 | |||
AK138817.1 | - | 2 | 290 | |||
BC004637.1 | - | 2 | 333 | 3586055 | ||
BC093528.1 | - | 2 | 607 | 5135606 | ||
BC172078.1 | - | 2 | 316 | 9054364 |