Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 2 | 454 | ||
NM_023210.4 | NP_075699.3 | 2 | 454 | |||
GenBank mRNA | AB037685.1 | - | 2 | 129 | ||
AK012759.1 | - | 2 | 421 | |||
AK049647.1 | - | 2 | 421 | |||
AK076049.1 | - | 2 | 281 | |||
AK088401.1 | - | 2 | 434 | |||
AK147888.1 | - | 2 | 420 | |||
AK168380.1 | - | 2 | 403 | |||
AK172146.1 | - | 2 | 468 | |||
AK205819.1 | - | 1 | 68 | |||
BC005690.1 | - | 2 | 389 | 3596597 | ||
BC080684.1 | - | 2 | 394 | 6493476 | ||
CF952181.1 | - | 2 | 125 | |||
U89345.2 | - | 2 | 415 |