Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023041.3 | NP_075528.3 | 3 | 248 | ||
GenBank mRNA | AK012785.1 | - | 3 | 231 | ||
AK029368.1 | - | 3 | 249 | |||
AK075576.1 | - | 3 | 235 | |||
AK170175.1 | - | 3 | 237 | |||
BC012517.1 | - | 3 | 183 | |||
BC019767.1 | - | 3 | 231 | 4502819 | ||
CT010305.1 | - | 3 | 224 | |||
Y09046.1 | - | 3 | 229 |