Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081682.2 | NP_001075151.1 | 4 | 462 | ||
NM_001285491.1 | NP_001272420.1 | 4 | 462 | |||
NM_001285493.1 | NP_001272422.1 | 4 | 404 | |||
NM_001285494.1 | NP_001272423.1 | 4 | 404 | |||
NM_023120.5 | NP_075609.2 | 4 | 404 | |||
GenBank mRNA | AF301595.1 | - | 4 | 355 | ||
AK010489.1 | - | 4 | 362 | |||
AK016952.1 | - | 4 | 392 | |||
AK087954.1 | - | 4 | 461 | |||
AK158210.1 | - | 4 | 391 | |||
AK166195.1 | - | 4 | 388 | |||
AK171253.1 | - | 4 | 384 | |||
AK173227.1 | - | 4 | 387 | |||
BC024635.1 | - | 4 | 373 | 4193427 | ||
BC037676.1 | - | 3 | 290 | 3493788 |
Set Membership: |
G-protein coupled receptor |