Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023173.2 | NP_075662.2 | 3 | 696 | ||
GenBank mRNA | AF268196.1 | - | 3 | 621 | ||
AF280810.1 | - | 3 | 563 | |||
AK004488.1 | - | 3 | 578 | |||
BC099453.1 | - | 3 | 696 | 30938295 |