Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159525.1 | NP_001152997.1 | 3, 4 | 335 | ||
NM_023041.3 | NP_075528.3 | 5, 6 | 611 | |||
GenBank mRNA | AK012785.1 | - | 5, 6 | 594 | ||
AK028449.1 | - | 3, 4 | 329 | |||
AK029368.1 | - | 5, 6 | 612 | |||
AK075576.1 | - | 5, 6 | 598 | |||
AK170175.1 | - | 5, 6 | 600 | |||
AK182815.1 | - | 2, 3 | 193 | |||
AK218946.1 | - | 2, 3 | 193 | |||
BC012517.1 | - | 5, 6 | 546 | |||
BC019767.1 | - | 5, 6 | 594 | 4502819 | ||
CT010305.1 | - | 5, 6 | 587 | |||
Y09046.1 | - | 5, 6 | 592 |