Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023049.1 | NP_075536.1 | 7, 8 | 1438 | ||
GenBank mRNA | AB221595.1 | - | 6, 7 | 1048 | ||
AF155353.1 | - | 5, 6 | 905 | |||
AK083400.1 | - | 5, 6 | 964 | |||
AK155235.1 | - | 3, 4 | 1152 | |||
BC031161.1 | - | 7, 8 | 1438 | 4981777 |