Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 5 | 952 | ||
NM_001164208.1 | NP_001157680.1 | 5 | 600 | |||
NM_001164209.2 | NP_001157681.1 | 6 | 808 | |||
NM_001286651.1 | NP_001273580.1 | 6 | 805 | |||
NM_001286652.1 | NP_001273581.1 | 5 | 712 | |||
NM_023056.4 | NP_075543.1 | 5 | 1201 | |||
GenBank mRNA | AB031386.1 | - | 5 | 761 | ||
AF115426.1 | - | 5 | 673 | |||
AK007408.1 | - | 5 | 607 | |||
AK156042.1 | - | 5 | 953 | |||
BC019539.1 | - | 4 | 570 | 4237639 | ||
BC022166.1 | - | 5 | 1216 | 3582123 | ||
BQ571258.1 | - | 5 | 592 | |||
CT010176.1 | - | 4 | 565 | |||
DV047496.1 | - | 5 | 639 | |||
DV059241.1 | - | 5 | 676 |