Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290381.1 | NP_001277310.1 | 2, 3 | 645 | ||
NM_023524.2 | NP_076013.1 | 2, 3 | 645 | |||
GenBank mRNA | AB035134.1 | - | 2, 3 | 336 | ||
AF267988.1 | - | 2, 3 | 600 | |||
AK010279.1 | - | 2, 3 | 303 | |||
AK155925.1 | - | 2, 3 | 629 | |||
BC024356.1 | - | 2, 3 | 544 | 5066740 | ||
CX768547.1 | - | 1, 2 | 164 |