Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7692
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
23
4740
NM_001256325.1
NP_001243254.1
23
4740
NM_001256326.1
NP_001243255.1
23
4740
NM_001256327.1
NP_001243256.1
23
4740
NM_001256328.1
NP_001243257.1
23
4740
NM_001256329.1
NP_001243258.1
23
4740
NM_001256330.1
NP_001243259.1
23
4740
NM_001256331.1
NP_001243260.1
23
4740
NM_001256332.1
NP_001243261.1
22
4671
NM_001256333.1
NP_001243262.1
23
4740
NM_001256334.1
NP_001243263.1
23
4740
NM_001256359.1
NP_001243288.1
23
4740
NM_001256360.1
NP_001243289.1
23
4740
NM_001256361.1
NP_001243290.1
23
4740
NM_018896.4
NP_061496.2
23
4740
NM_198376.2
NP_938190.1
22
4671
NM_198377.2
NP_938191.2
23
4740
NM_198378.2
NP_938192.1
23
4740
NM_198379.2
NP_938193.1
22
4671
NM_198380.2
NP_938194.1
23
4740
NM_198382.2
NP_938196.1
22
4671
NM_198383.2
NP_938197.1
22
4671
NM_198384.2
NP_938198.1
23
4740
NM_198385.2
NP_938199.1
23
4740
NM_198386.2
NP_938200.1
23
4740
NM_198387.2
NP_938201.1
22
4671
NM_198388.2
NP_938202.1
22
4671
NM_198396.2
NP_938406.1
22
4671
NR_046054.1
23
4740
NR_046055.1
23
4740
NR_046056.1
23
4740
NR_046057.1
22
4671
NR_046058.1
22
4671
XM_006722160.3
XP_006722223.1
22
4810
XM_006722161.3
XP_006722224.1
22
4759
GenBank mRNA
AB012043.1
-
23
4376
AB032949.2
-
16
2702
AF126965.1
-
22
4454
AF126966.1
-
22
4454
AF134985.1
-
22
4279
AF134986.1
-
23
4348
AF190860.1
-
22
4495
AF227744.1
-
23
4348
AF227745.1
-
23
4348
AF227746.1
-
22
4279
AF227747.1
-
22
4279
AF227748.1
-
22
4279
AF227749.1
-
23
4348
AF227750.1
-
23
4348
AF227751.1
-
23
4348
AK296513.1
-
10
1451
BC110995.1
-
22
4671
5502027
DQ494449.1
-
22
4279
DQ494450.1
-
22
4279
DQ494451.1
-
22
4279
DQ494452.1
-
22
4279
DQ494453.1
-
22
4279
DQ494454.1
-
22
4279
DQ494455.1
-
22
4279
DQ494456.1
-
22
4279
DQ494457.1
-
23
4348
DQ494458.1
-
23
4348
DQ494459.1
-
23
4348
DQ494460.1
-
23
4348
DQ494461.1
-
23
4348
DQ494462.1
-
23
4348
DQ494463.1
-
23
4348
DQ494464.1
-
23
4348
DQ494465.1
-
23
4348
DQ494466.1
-
23
4348
DQ494467.1
-
23
4348
DQ494468.1
-
23
4348
DQ494469.1
-
23
4348
DQ494470.1
-
23
4348
DQ494471.1
-
23
4348
DQ494472.1
-
22
4279
DQ494473.1
-
23
4348
DQ494474.1
-
23
4348
DQ494475.1
-
23
4348
DQ494476.1
-
23
4348
DQ494477.1
-
23
4348
DQ494478.1
-
23
4348
DQ494479.1
-
23
4348
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0