Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048008.2 | NP_001041473.1 | 8, 9 | 829 | ||
NM_001284218.1 | NP_001271147.1 | 8, 9 | 954 | |||
NM_001284221.1 | NP_001271150.1 | 7, 8 | 704 | |||
NM_023328.3 | NP_075817.2 | 8, 9 | 829 | |||
GenBank mRNA | AF219141.1 | - | 8, 9 | 768 | ||
AK015394.1 | - | 7, 8 | 668 | |||
AK132695.1 | - | 8, 9 | 769 | |||
AK133215.1 | - | 8, 9 | 888 | |||
AK170046.1 | - | 8, 9 | 772 | |||
BC060633.1 | - | 8, 9 | 873 | |||
BC082335.1 | - | 7, 8 | 692 | 30536788 | ||
CN527234.1 | - | 5, 6 | 515 |