Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254761.1 | NP_001241690.1 | 3 | 812 | ||
NM_023270.5 | NP_075759.3 | 3 | 938 | |||
GenBank mRNA | AB041548.1 | - | 3 | 938 | ||
AF426411.1 | - | 3 | 752 | |||
AK004847.1 | - | 3 | 939 | |||
AK018582.1 | - | 3 | 940 | |||
AK146266.1 | - | 3 | 935 | |||
AK167031.1 | - | 3 | 924 | |||
AY112656.1 | - | 3 | 925 | |||
BC010477.1 | - | 3 | 927 | 3154433 | ||
DV064262.1 | - | 3 | 575 |