Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023844.5 | NP_076333.3 | 2 | 447 | ||
GenBank mRNA | AF255911.1 | - | 2 | 170 | ||
AJ291757.1 | - | 2 | 383 | |||
AK010616.1 | - | 2 | 289 | |||
AK010826.1 | - | 2 | 291 | |||
AK013914.1 | - | 2 | 288 | |||
AK028757.1 | - | 2 | 138 | |||
AK045095.1 | - | 2 | 185 | |||
AK078128.1 | - | 2 | 173 | |||
AK153636.1 | - | 2 | 136 | |||
BC028778.1 | - | 2 | 447 | 1195543 |