Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023603.3 | NP_076092.1 | 2 | 960 | ||
GenBank mRNA | AF272847.1 | - | 2 | 253 | ||
AK157783.1 | - | 3 | 597 | |||
AY034062.1 | - | 2 | 875 | |||
BC080848.1 | - | 2 | 926 | |||
BC089305.1 | - | 2 | 926 | 30360144 |