Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023734.3 | NP_076223.3 | 2 | 671 | ||
GenBank mRNA | AB046537.1 | - | 320 | |||
AK004672.1 | - | 2 | 365 | |||
AK157854.1 | - | 2 | 677 | |||
AK158339.1 | - | 2 | 673 | |||
BC039124.1 | - | 2 | 268 | |||
BC106088.1 | - | 2 | 356 | |||
BC118026.1 | - | 2 | 319 |