Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205385.1 | NP_001192314.1 | 5 | 487 | ||
NM_001205386.1 | NP_001192315.1 | 5 | 484 | |||
NM_023735.2 | NP_076224.1 | 4 | 679 | |||
GenBank mRNA | AK004554.1 | - | 4 | 493 | ||
AK083343.1 | - | 4 | 497 | |||
AK145536.1 | - | 4 | 499 | |||
AK147931.1 | - | 4 | 329 | |||
AK150487.1 | - | 4 | 381 | |||
AK150589.1 | - | 4 | 457 | |||
AK150991.1 | - | 4 | 437 | |||
AK152865.1 | - | 4 | 474 | |||
AK153079.1 | - | 4 | 500 | |||
AK153150.1 | - | 4 | 474 | |||
AK153204.1 | - | 4 | 380 | |||
AK159135.1 | - | 4 | 466 | |||
AK159668.1 | - | 4 | 391 | |||
AK159775.1 | - | 4 | 478 | |||
AK160047.1 | - | 4 | 339 | |||
AK166529.1 | - | 5 | 431 | |||
AK166586.1 | - | 5 | 431 | |||
AK167148.1 | - | 4 | 438 | |||
AK167891.1 | - | 4 | 436 | |||
AK168568.1 | - | 4 | 366 | |||
AK168771.1 | - | 4 | 452 | |||
BC005557.1 | - | 4 | 442 | 3156748 | ||
BC080806.1 | - | 5 | 445 | 5350752 | ||
BC108386.1 | - | 4 | 410 | 5051125 |