Gene Symbol: Ndufc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024220.2 | NP_077182.1 | 2 | 452 | ||
GenBank mRNA | AK002373.1 | - | 2 | 360 | ||
AK003380.2 | - | 2 | 355 | |||
AK008502.1 | - | 2 | 367 | |||
AK008889.1 | - | 2 | 360 | |||
AK019002.1 | - | 2 | 379 | |||
AK153547.1 | - | 2 | 382 | |||
AK166594.1 | - | 2 | 379 | |||
AK168342.1 | - | 2 | 404 | |||
BC002097.1 | - | 2 | 375 | 3497384 | ||
CK128102.1 | - | 2 | 452 |