Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128214.1 | NP_001121686.1 | 3 | 647 | ||
NM_153331.3 | NP_699162.3 | 2 | 921 | |||
XM_005264937.2 | XP_005264994.1 | 3 | 1979 | |||
GenBank mRNA | AK027572.1 | - | 2 | 1513 | ||
AK074934.1 | - | 2 | 921 | |||
AK075297.1 | - | 3 | 629 | |||
BC022893.1 | - | 2 | 663 | 4689885 |