Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024190.2 | NP_077152.1 | 1 | 276 | ||
GenBank mRNA | AK012996.1 | - | 1 | 226 | ||
AK014091.1 | - | 1 | 243 | |||
AK151071.1 | - | 1 | 245 | |||
AK167614.1 | - | 1 | 214 | |||
BC002229.1 | - | 1 | 212 | 3491354 | ||
BC034442.1 | - | 1 | 200 | |||
CT010307.1 | - | 1 | 134 |