Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024209.2 | NP_077171.1 | 3 | 310 | ||
GenBank mRNA | AK002764.1 | - | 3 | 272 | ||
AK009104.1 | - | 3 | 290 | |||
BC002223.1 | - | 3 | 335 | 3491111 |