Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024215.2 | NP_077177.2 | 2, 3 | 359 | ||
GenBank mRNA | AK005247.1 | - | 2, 3 | 361 | ||
AK133585.1 | - | 2, 3 | 357 | |||
AK156931.1 | - | 2, 3 | 294 | |||
BC003465.1 | - | 2, 3 | 296 | 2648747 |