Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024217.3 | NP_077179.1 | 5 | 1127 | ||
GenBank mRNA | AK003882.1 | - | 5 | 983 | ||
AK079165.1 | - | 5 | 1096 | |||
AK172091.1 | - | 5 | 1030 | |||
AK198392.1 | - | 1 | 222 | |||
AY241870.1 | - | 5 | 1121 | |||
BC003230.1 | - | 5 | 1034 | 3483351 |