Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 10 | 1103 | ||
GenBank mRNA | AK011810.1 | - | 1067 | |||
AK014180.1 | - | 10 | 1058 | |||
AK014918.1 | - | 10 | 1102 | |||
AK153058.1 | - | 10 | 1057 | |||
AK166631.1 | - | 10 | 1105 | |||
AK167798.1 | - | 10 | 1059 | |||
BC002188.1 | - | 8 | 927 | |||
BC019512.1 | - | 10 | 1076 | 4210204 | ||
BC094468.1 | - | 10 | 1049 | 5011266 |