Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024246.5 | NP_077208.1 | 4 | 1636 | ||
GenBank mRNA | AK009760.1 | - | 4 | 1637 | ||
AK010144.1 | - | 4 | 1726 | |||
BC003309.1 | - | 4 | 1691 | 3154669 |